accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 412947-54-7

IUPAC Name: methyl 3-amino-4-iodobenzoate
Synonyms: methyl 3-amino-4-iodobenzoate
Molecular Weight (g/mol): 277.06
Molecular Formula: C8H8INO2
InChi Key: WJEBNIVVLJEIKE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(N)=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results