accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41295-20-9

IUPAC Name: 4-(4-methylphenoxy)aniline
Synonyms: 4-(4-methylphenoxy)aniline
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: VPCGOYHSWIYEMO-UHFFFAOYSA-N
SMILES: CC1=CC=C(OC2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results