accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41295-98-1

NaNH2ClClSOOO

CAS RN 41295-98-1

IUPAC Name: sodium 4-amino-2,5-dichlorobenzene-1-sulfonate
Synonyms: sodium 4-amino-2,5-dichlorobenzenesulfonate
Molecular Weight (g/mol): 264.05
Molecular Formula: C6H4Cl2NNaO3S
InChi Key: MOJYHTGMAPQZMC-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC(Cl)=C(C=C1Cl)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)