accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4132-48-3

IUPAC Name: 1-methoxy-4-(propan-2-yl)benzene
Synonyms: 1-isopropyl-4-methoxybenzene
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: JULZQKLZSNOEEJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results