accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOOCH3

CAS RN 41351-30-8

IUPAC Name: 4-(2,4-dimethoxybenzoyl)phenol
Synonyms: 4-(2,4-dimethoxybenzoyl)phenol
Molecular Weight (g/mol): 258.27
Molecular Formula: C15H14O4
InChi Key: QEHRETCJMLQPCR-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C=C1)C(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)