accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClMgBr

CAS RN 413589-34-1

IUPAC Name: bromo(3-chloro-4-fluorophenyl)magnesium
Synonyms: bromo(3-chloro-4-fluorophenyl)magnesium
Molecular Weight (g/mol): 233.75
Molecular Formula: C6H3BrClFMg
InChi Key: XRNCDMQVKMYTAM-UHFFFAOYSA-M
SMILES: FC1=CC=C([Mg]Br)C=C1Cl
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results