accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4136-91-8

IUPAC Name: N,N-bis(propan-2-yl){[bis(propan-2-yl)carbamothioyl]disulfanyl}carbothioamide
Synonyms: N,N-diisopropyl(diisopropylcarbamothioyl)disulfanylcarbothioamide
Molecular Weight (g/mol): 352.63
Molecular Formula: C14H28N2S4
InChi Key: ZUYREEAWHZRZDX-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=S)SSC(=S)N(C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results