accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OCH3(S)H2NOHHOOHOCH3(S)H2NOHHOOHO

CAS RN 41372-08-1

IUPAC Name: bis((2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid) trihydrate
Synonyms: bis(methyldopa) trihydrate
Molecular Weight (g/mol): 476.48
Molecular Formula: C20H32N2O11
InChi Key: YKFCISHFRZHKHY-NGQGLHOPSA-N
SMILES: O.O.O.C[C@](N)(CC1=CC=C(O)C(O)=C1)C(O)=O.C[C@](N)(CC1=CC=C(O)C(O)=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)