accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41381-89-9

IUPAC Name: 2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
Synonyms: 2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
Molecular Weight (g/mol): 200.26
Molecular Formula: C11H8N2S
InChi Key: SOFCSIBVJROWGI-UHFFFAOYSA-N
SMILES: N#CCC1=NC(=CS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results