accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOO

CAS RN 4143-64-0

IUPAC Name: 2-(3,4-dihydroxyphenyl)-4H-chromen-4-one
Synonyms: 2-(3,4-dihydroxyphenyl)chromen-4-one
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: SRNPMQHYWVKBAV-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1O)C1=CC(=O)C2=CC=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results