accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NNHCH3NOSOONH3CH3C

CAS RN 41483-43-6

IUPAC Name: 5-butyl-2-(ethylamino)-6-methylpyrimidin-4-yl N,N-dimethylsulfamate
Synonyms: bupirimate
Molecular Weight (g/mol): 316.42
Molecular Formula: C13H24N4O3S
InChi Key: DSKJPMWIHSOYEA-UHFFFAOYSA-N
SMILES: CCCCC1=C(C)N=C(NCC)N=C1OS(=O)(=O)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results