accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNO

CAS RN 4149-06-8

IUPAC Name: 3-amino-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 5-amino-2-phenyl-4H-pyrazol-3-one
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: LPOVZHYARSAVIZ-UHFFFAOYSA-N
SMILES: NC1=NN(C(=O)C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results