accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41513-04-6

IUPAC Name: 1-bromo-4-chloro-2-nitrobenzene
Synonyms: 1-bromo-4-chloro-2-nitrobenzene
Molecular Weight (g/mol): 236.45
Molecular Formula: C6H3BrClNO2
InChi Key: UKTIMFAJRPSNGR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results