accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3OOCH3(E)

CAS RN 41530-32-9

IUPAC Name: 1,3-dimethyl 2-[(2E)-3-methoxyprop-2-en-1-ylidene]propanedioate
Synonyms: 1,3-dimethyl 2-[(2E)-3-methoxyprop-2-en-1-ylidene]propanedioate
Molecular Weight (g/mol): 200.19
Molecular Formula: C9H12O5
InChi Key: NOBNTZJTOBIOHL-GQCTYLIASA-N
SMILES: CO\C=C\C=C(C(=O)OC)C(=O)OC
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results