CAS RN 41668-13-7
IUPAC Name:
5-bromo-6-oxo-1,6-dihydropyridine-3-carboxylic acid
Synonyms:
5-bromo-6-oxo-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol):
218.01
Molecular Formula:
C6H4BrNO3
InChi Key:
PQDLYKZCJBGXPQ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CNC(=O)C(Br)=C1
1
–
1
of
1
results
1
5-Bromo-6-hydroxynicotinic Acid 98.0+%, TCI America™
CAS: 41668-13-7 Molecular Formula: C6H4BrNO3 Molecular Weight (g/mol): 218.006 MDL Number: MFCD08235173 InChI Key: PQDLYKZCJBGXPQ-UHFFFAOYSA-N Synonym: 5-Bromo-6-hydroxy-3-pyridinecarboxylic Acid PubChem CID: 2764314 IUPAC Name: 5-bromo-6-oxo-1H-pyridine-3-carboxylic acid SMILES: C1=C(C(=O)NC=C1C(=O)O)Br