accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4171-83-9

ONOS

CAS RN 4171-83-9

IUPAC Name: 1-nitro-2-(phenylsulfanyl)benzene
Synonyms: 1-nitro-2-(phenylsulfanyl)benzene
Molecular Weight (g/mol): 231.27
Molecular Formula: C12H9NO2S
InChi Key: ZPWNCSAEXUDWTN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1SC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)