CAS RN 41762-76-9
IUPAC Name:
3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl chloride
Synonyms:
3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl chloride
Molecular Weight (g/mol):
226.63
Molecular Formula:
C5H7ClN2O4S
InChi Key:
ZWTPALHHEULAPI-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)N1CCN(C(Cl)=O)C1=O