accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNN

CAS RN 4177-16-6

IUPAC Name: 2-ethenylpyrazine
Synonyms: 2-ethenylpyrazine
Molecular Weight (g/mol): 106.13
Molecular Formula: C6H6N2
InChi Key: KANZWHBYRHQMKZ-UHFFFAOYSA-N
SMILES: C=CC1=CN=CC=N1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)