accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3NOH

CAS RN 41772-23-0

IUPAC Name: 1-hydroxynaphthalen-2-aminium chloride
Synonyms: 1-hydroxynaphthalen-2-aminium chloride
Molecular Weight (g/mol): 195.65
Molecular Formula: C10H10ClNO
InChi Key: PEJOQASNBCUDMB-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=CC=C2C=CC=CC2=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results