accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)NH2ONHNOO

CAS RN 4178-93-2

IUPAC Name: (2S)-2-amino-4-methyl-N-(4-nitrophenyl)pentanamide
Synonyms: (2S)-2-amino-4-methyl-N-(4-nitrophenyl)pentanamide
Molecular Weight (g/mol): 251.29
Molecular Formula: C12H17N3O3
InChi Key: AXZJHDNQDSVIDR-NSHDSACASA-N
SMILES: CC(C)C[C@H](N)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)