CAS RN 4182-44-9
IUPAC Name:
bis(2-aminoethyl)(dodecyl)amine
Synonyms:
bis(2-aminoethyl)(dodecyl)amine
Molecular Weight (g/mol):
271.49
Molecular Formula:
C16H37N3
InChi Key:
BRNPMBFYMPUNSK-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCN(CCN)CCN
1
–
1
of
1
results
1
4-Dodecyldiethylenetriamine (Tech.), 90%, Thermo Scientific™
CAS: 4182-44-9 Molecular Formula: C16H37N3 Molecular Weight (g/mol): 271.493 InChI Key: BRNPMBFYMPUNSK-UHFFFAOYSA-N Synonym: 4-dodecyldiethylenetriamine,unii-vg94uin7ys,1,2-ethanediamine, n-2-aminoethyl-n-dodecyl,n-2-aminoethyl-n-dodecyl-1,2-ethanediamine,vg94uin7ys,n1-2-aminoethyl-n1-dodecyl-1,2-ethanediamine,1,2-ethanediamine, n1-2-aminoethyl-n1-dodecyl,n-2-aminoethyl-n-dodecylethylenediamine,bis 2-aminoethyl dodecyl amine,n'-2-aminoethyl-n'-dodecylethane-1,2-diamine PubChem CID: 77847 IUPAC Name: N'-(2-aminoethyl)-N'-dodecylethane-1,2-diamine SMILES: CCCCCCCCCCCCN(CCN)CCN