accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3NH2

CAS RN 41851-59-6

IUPAC Name: 1-(4-methoxyphenyl)ethan-1-amine
Synonyms: 1-(4-methoxyphenyl)ethanamine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: JTDGKQNNPKXKII-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(C)N
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results