CAS RN 41864-24-8
CAS RN 41864-24-8
IUPAC Name:
tert-butyl 1H-1,2,4-triazole-1-carboxylate
Synonyms:
tert-butyl 1,2,4-triazole-1-carboxylate
Molecular Weight (g/mol):
169.18
Molecular Formula:
C7H11N3O2
InChi Key:
XWUFRDJEUOAMNW-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1C=NC=N1
1
–
1
of
1
results
1
1-tert-Butoxycarbonyl-1,2,4-triazole 97.0+%, TCI America™
CAS: 41864-24-8 Molecular Formula: C7H11N3O2 Molecular Weight (g/mol): 169.18 MDL Number: MFCD00042665 InChI Key: XWUFRDJEUOAMNW-UHFFFAOYSA-N Synonym: 1-Boc-1,2,4-triazole PubChem CID: 3716936 IUPAC Name: tert-butyl 1H-1,2,4-triazole-1-carboxylate SMILES: CC(C)(C)OC(=O)N1C=NC=N1