accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2NH2CH3

CAS RN 41927-01-9

IUPAC Name: 3,4-dimethylbenzene-1,2-diamine
Synonyms: 3,4-dimethylbenzene-1,2-diamine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: MHQULXYNBKWNDF-UHFFFAOYSA-N
SMILES: CC1=C(C)C(N)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)