CAS RN 41931-13-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ SOD1 Inhibitor, LCS-1
The SOD1 Inhibitor, LCS-1, also referenced under CAS 41931-13-9, controls the biological activity of SOD1.