accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41934-47-8

IUPAC Name: 7-(diethylamino)-4-(trifluoromethyl)-2H-chromen-2-one
Synonyms: coumarin 481 coumarin 152A
Molecular Weight (g/mol): 285.27
Molecular Formula: C14H14F3NO2
InChi Key: UIMOXRDVWDLOHW-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results