accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 419535-75-4

IUPAC Name: bromo(5-chloro-2-methoxyphenyl)magnesium
Synonyms: bromo(5-chloro-2-methoxyphenyl)magnesium
Molecular Weight (g/mol): 245.78
Molecular Formula: C7H6BrClMgO
InChi Key: GAFVLSUAFZZUMW-UHFFFAOYSA-M
SMILES: COC1=CC=C(Cl)C=C1[Mg]Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results