accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrN

CAS RN 41963-20-6

IUPAC Name: 4-bromo-3-methylbenzonitrile
Synonyms: 4-bromo-3-methylbenzonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: SKXUZFJOLNNWIG-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1Br)C#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)