accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHON

CAS RN 4198-90-7

IUPAC Name: 4-hydroxy-3,5-dimethylbenzonitrile
Synonyms: 4-hydroxy-3,5-dimethylbenzonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: WFYGXOWFEIOHCZ-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1O)C#N
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)