accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONN

CAS RN 4199-88-6

IUPAC Name: 5-nitro-1,10-phenanthroline
Synonyms: 1,10-phenanthroline, 5-nitro-
Molecular Weight (g/mol): 225.21
Molecular Formula: C12H7N3O2
InChi Key: PDDBTWXLNJNICS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=CC=CN=C2C2=NC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)