accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 4200-92-4

IUPAC Name: 3-octyloxolane-2,5-dione
Synonyms: 3-octyloxolane-2,5-dione
Molecular Weight (g/mol): 212.29
Molecular Formula: C12H20O3
InChi Key: OAJCSERLBQROJC-UHFFFAOYNA-N
SMILES: CCCCCCCCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)