accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOHO

CAS RN 42013-20-7

IUPAC Name: 2-[(2-methylphenyl)formamido]acetic acid
Synonyms: methyl hippurate
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: YOEBAVRJHRCKRE-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(=O)NCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)