accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 420137-14-0

IUPAC Name: 3-(benzyloxy)pyrrolidine
Synonyms: 3-(benzyloxy)pyrrolidine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: CWBMYKUPMLRKQK-UHFFFAOYNA-N
SMILES: C(OC1CCNC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results