accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOONOO

CAS RN 42014-51-7

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-bromoacetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-bromoacetate
Molecular Weight (g/mol): 236.02
Molecular Formula: C6H6BrNO4
InChi Key: NKUZQMZWTZAPSN-UHFFFAOYSA-N
SMILES: BrCC(=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)