accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 4203-50-3

IUPAC Name: 2-phenoxyoxane
Synonyms: 2-phenoxyoxane
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: SHRDVLUJLDYXSO-UHFFFAOYNA-N
SMILES: C1CCC(OC2=CC=CC=C2)OC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results