CAS RN 42057-22-7
IUPAC Name:
(2S,5R,6R)-6-[(2R)-2-[(3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium
Synonyms:
mezlocillin sodium
Molecular Weight (g/mol):
562.57
Molecular Formula:
C21H25N5NaO8S2
InChi Key:
SGVORSZSYKDVFT-ZBJAFUORSA-N
SMILES:
[Na].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](NC(=O)N3CCN(C3=O)S(C)(=O)=O)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Mezlocillin sodium, 97%
CAS: 42057-22-7 Molecular Formula: C21H24N5NaO8S2 Molecular Weight (g/mol): 561.56 InChI Key: BXRKBOLIXGCTKD-SYNJJEHYSA-N PubChem CID: 131673974 IUPAC Name: (2S,5R,6R)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium SMILES: [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]