accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3CI

CAS RN 4206-67-1

IUPAC Name: (iodomethyl)trimethylsilane
Synonyms: (iodomethyl)trimethylsilane
Molecular Weight (g/mol): 214.12
Molecular Formula: C4H11ISi
InChi Key: VZNYXGQMDSRJAL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CI
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results