accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42066-86-4

IUPAC Name: 2-isocyanatobenzonitrile
Synonyms: 2-isocyanatobenzonitrile
Molecular Weight (g/mol): 144.13
Molecular Formula: C8H4N2O
InChi Key: FQLNVWAFXQUXGW-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results