accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOONOOOOH

CAS RN 420841-84-5

IUPAC Name: 2-[3-(4-nitrophenoxy)phenyl]-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid
Synonyms: 2-[3-(4-nitrophenoxy)phenyl]-1,3-dioxoisoindole-5-carboxylic acid
Molecular Weight (g/mol): 404.33
Molecular Formula: C21H12N2O7
InChi Key: HFYPTENHTPNXGP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(C=C1)C(=O)N(C2=O)C1=CC(OC2=CC=C(C=C2)[N+]([O-])=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results