accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4209-18-1

IUPAC Name: 3-benzyl-4-methyl-1,3-thiazol-3-ium chloride
Synonyms: 3-benzyl-4-methyl-1,3-thiazol-3-ium chloride
Molecular Weight (g/mol): 225.73
Molecular Formula: C11H12ClNS
InChi Key: ZGBNEPNQIDHABT-UHFFFAOYSA-M
SMILES: [Cl-].CC1=CSC=[N+]1CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results