CAS RN 42097-42-7
IUPAC Name:
2-(hydroxymethyl)-6-methylpyridin-3-ol
Synonyms:
2-(hydroxymethyl)-6-methylpyridin-3-ol
Molecular Weight (g/mol):
139.15
Molecular Formula:
C7H9NO2
InChi Key:
PAGTXDLKXRBHFL-UHFFFAOYSA-N
SMILES:
CC1=CC=C(O)C(CO)=N1
1
–
1
of
1
results
1
Sigma Aldrich 2,6-Lutidine-alpha2,3-diol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.