accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 421-70-5

IUPAC Name: 1-bromo-1,1,2,2-tetrafluoro-2-iodoethane
Synonyms: 1-bromo-1,1,2,2-tetrafluoro-2-iodoethane
Molecular Weight (g/mol): 306.82
Molecular Formula: C2BrF4I
InChi Key: ZYNPYKGTNSXKPI-UHFFFAOYSA-N
SMILES: FC(F)(Br)C(F)(F)I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)