accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NN

CAS RN 4210-60-0

IUPAC Name: 2-tert-butylpropanedinitrile
Synonyms: 2-tert-butylpropanedinitrile
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: TUJPHMQZSDKRSL-UHFFFAOYSA-N
SMILES: CC(C)(C)C(C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results