accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42104-58-5

IUPAC Name: {[2-(phosphonomethyl)phenyl]methyl}phosphonic acid
Synonyms: [2-(phosphonomethyl)phenyl]methylphosphonic acid
Molecular Weight (g/mol): 266.13
Molecular Formula: C8H12O6P2
InChi Key: ZCVUWZJGYKGTPG-UHFFFAOYSA-N
SMILES: OP(O)(=O)CC1=CC=CC=C1CP(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results