CAS RN 42122-75-8
IUPAC Name:
methyl 5-amino-2-chlorobenzoate
Synonyms:
methyl 5-amino-2-chlorobenzoate
Molecular Weight (g/mol):
185.61
Molecular Formula:
C8H8ClNO2
InChi Key:
LBNPBOFVHYOPIB-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(Cl)C=CC(N)=C1
1
–
1
of
1
results
1
Methyl 5-Amino-2-chlorobenzoate 98.0+%, TCI America™
CAS: 42122-75-8 Molecular Formula: C8H8ClNO2 Molecular Weight (g/mol): 185.61 MDL Number: MFCD07800315 InChI Key: LBNPBOFVHYOPIB-UHFFFAOYSA-N Synonym: 5-Amino-2-chlorobenzoic Acid Methyl Ester PubChem CID: 5200412 IUPAC Name: methyl 5-amino-2-chlorobenzoate SMILES: COC(=O)C1=C(Cl)C=CC(N)=C1