accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42134-49-6

IUPAC Name: triphenyl[3-(trimethylsilyl)prop-2-yn-1-yl]phosphanium bromide
Synonyms: triphenyl[3-(trimethylsilyl)prop-2-yn-1-yl]phosphanium bromide
Molecular Weight (g/mol): 453.44
Molecular Formula: C24H26BrPSi
InChi Key: PBSHVEOONSKWJF-UHFFFAOYSA-M
SMILES: [Br-].C[Si](C)(C)C#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results