accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNOO

CAS RN 42137-23-5

IUPAC Name: 5-nitrothiophene-3-carbonitrile
Synonyms: 5-nitrothiophene-3-carbonitrile
Molecular Weight (g/mol): 154.14
Molecular Formula: C5H2N2O2S
InChi Key: NACMZLGGHMPMKQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CS1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results