accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 421552-12-7

IUPAC Name: 2-(bromomethyl)-4-fluorobenzonitrile
Synonyms: 2-(bromomethyl)-4-fluorobenzonitrile
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: CHCAGFNTASDQFX-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results