accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOHNH2OCH3(E/Z)

CAS RN 42191-48-0

IUPAC Name: 2-(3,4-dimethoxyphenyl)-N'-hydroxyethanimidamide
Synonyms: 2-(3,4-dimethoxyphenyl)-N'-hydroxyethanimidamide
Molecular Weight (g/mol): 210.23
Molecular Formula: C10H14N2O3
InChi Key: WJFPRBYVVFXVFB-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(CC(N)=NO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results