accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42196-31-6

IUPAC Name: palladium(2+) ditrifluoroacetate
Synonyms: palladium(2+) ditrifluoroacetate
Molecular Weight (g/mol): 332.45
Molecular Formula: C4F6O4Pd
InChi Key: PBDBXAQKXCXZCJ-UHFFFAOYSA-L
SMILES: [Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)